AMBH2D6EDCE1 Ambeed, Inc.

Cyproheptadine Hydrochloride Sesquihydrate

  • MDL number MFCD27967225
  • Cas Number : 41354-29-4
  • Molecular Weight : 701.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/41354-29-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/41354-29-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart