AMBH2D6EDD78 Ambeed, Inc.

Octyl 2-aminoacetate hydrochloride

  • MDL number MFCD00151827
  • Cas Number : 39540-30-2
  • Molecular Weight : 223.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-72
Purity 97
Flash Point 108.5
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 243.5�t 760 mmHg
Product Link https://www.ambeed.com/products/39540-30-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/39540-30-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart