AMBH2D6EDDE3 Ambeed, Inc.

Dimethoxy(methyl)(phenyl)silane

  • MDL number MFCD00008343
  • Cas Number : 3027-21-2
  • Molecular Weight : 182.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3027-21-2.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/3027-21-2.html
pictograms GHS07
hstatements H227-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart