AMBH2D6EDE07 Ambeed, Inc.

Diphenyl(o-tolyl)phosphine

  • MDL number MFCD00008515
  • Cas Number : 5931-53-3
  • Molecular Weight : 276.3100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67-68
Purity 97
Flash Point 192.2
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Boiling point 378.3�t 760 mmHg
Product Link https://www.ambeed.com/products/5931-53-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5931-53-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart