AMBH2D6EDF86 Ambeed, Inc.

Ethyl 4-Methylbenzoylformate

  • MDL number MFCD01311169
  • Cas Number : 5524-56-1
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Flash Point 128.3°C
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 291.4°C at 760 mmHg
Product Link https://www.ambeed.com/products/5524-56-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5524-56-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart