AMBH2D6EEAA3 Ambeed, Inc.

5-(3-(Dimethylamino)propyl)phenanthridin-6(5H)-one

  • MDL number MFCD00865434
  • Cas Number : 17692-37-4
  • Molecular Weight : 280.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/17692-37-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/17692-37-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart