AMBH2D6EEE3A Ambeed, Inc.

Hematoporphyrin dihydrochloride

  • MDL number MFCD00013470
  • Cas Number : 17696-69-4
  • Molecular Weight : 671.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/17696-69-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/17696-69-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart