AMBH2D6EEE59 Ambeed, Inc.

Hoechst 33258 Trihydrochloride

  • MDL number MFCD00012679
  • Cas Number : 23491-45-4
  • Molecular Weight : 533.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(6-(4-Methylpiperazin-1-yl)-1H,3'H-[2,5'-bibenzo[d]imidazol]-2'-yl)phenol trihydrochloride
Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/23491-45-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/23491-45-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart