AMBH2D6EEED3 Ambeed, Inc.

6-Methylheptyl palmitate

  • MDL number MFCD00152588
  • Cas Number : 1341-38-4
  • Molecular Weight : 368.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 203.7
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 407.2�t 760 mmHg
Product Link https://www.ambeed.com/products/1341-38-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1341-38-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart