AMBH2D6EEF34 Ambeed, Inc.

Lodoxamide tromethamine

  • MDL number MFCD04112765
  • Cas Number : 63610-09-3
  • Molecular Weight : 553.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/63610-09-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/63610-09-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart