AMBH2D6EEF36 Ambeed, Inc.

Lomefloxacin hydrochloride

  • MDL number MFCD00214312
  • Cas Number : 98079-52-8
  • Molecular Weight : 387.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/98079-52-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/98079-52-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart