AMBH2D6EEF56 Ambeed, Inc.

2-Naphthamide

  • MDL number MFCD00016812
  • Cas Number : 2243-82-5
  • Molecular Weight : 171.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2243-82-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2243-82-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart