AMBH2D6EEF59 Ambeed, Inc.

Diisopropyl 2-(1,3-dithiol-2-ylidene)malonate

  • MDL number MFCD00867646
  • Cas Number : 59937-28-9
  • Molecular Weight : 288.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 321.5�t 760 mmHg
Iupac Name diisopropyl 2-(1,3-dithiol-2-ylidene)malonate
Inchi Code 1S/C12H16O4S2/c1-7(2)15-10(13)9(11(14)16-8(3)4)12-17-5-6-18-12/h5-8H,1-4H3
Inchi Key YPIQVCUJEKAZCP-UHFFFAOYSA-N

Description

Synonym Diisopropyl 2-(1,3-dithiol-2-ylidene)malonate
Purity 98%
Flash Point 138.7
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 321.5�t 760 mmHg
Product Link https://www.ambeed.com/products/59937-28-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/59937-28-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart