AMBH2D6EEF5A Ambeed, Inc.

Maltol propionate

  • MDL number MFCD00036605
  • Cas Number : 68555-63-5
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methyl-4-oxo-4H-pyran-3-yl propionate
Purity 97
Flash Point 141.5
Storage temp. Sealed in dry,Room Temperature
Physical Form Colorless to light yellow liquid or white to light yellow solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 317.8�t 760 mmHg
Product Link https://www.ambeed.com/products/68555-63-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/68555-63-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart