AMBH2D6EF065 Ambeed, Inc.

Methyl 2-oxoindoline-7-carboxylate

  • MDL number MFCD02179610
  • Cas Number : 380427-39-4
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-184
Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 402.7�t 760 mmHg
Product Link https://www.ambeed.com/products/380427-39-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/380427-39-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart