AMBH2D6EF0A1 Ambeed, Inc.

Mitoxantrone

  • MDL number MFCD00242942
  • Cas Number : 65271-80-9
  • Molecular Weight : 444.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-174
Synonym 1,4-Dihydroxy-5,8-bis((2-((2-hydroxyethyl)amino)ethyl)amino)anthracene-9,10-dione
Purity 98
Flash Point 441.1
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 805.7�t 760 mmHg
Product Link https://www.ambeed.com/products/65271-80-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/65271-80-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart