AMBH2D6EF0CA Ambeed, Inc.

H-Glu-Obzl

  • MDL number MFCD00037312
  • Cas Number : 13030-09-6
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157
Synonym (S)-4-Amino-5-(benzyloxy)-5-oxopentanoic acid
Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 416�t 760 mmHg
Product Link https://www.ambeed.com/products/13030-09-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13030-09-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart