AMBH2D6EF0D5 Ambeed, Inc.

4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane

  • MDL number MFCD02179447
  • Cas Number : 190788-59-1
  • Molecular Weight : 249.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/190788-59-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/190788-59-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart