AMBH2D6EF0EA Ambeed, Inc.

2-(Cyclohexylthio)isoindoline-1,3-dione

  • MDL number MFCD00072247
  • Cas Number : 17796-82-6
  • Molecular Weight : 261.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/17796-82-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/17796-82-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart