AMBH2D6EF4AD Ambeed, Inc.

Ac-DL-Abu-OH

  • MDL number MFCD00020430
  • Cas Number : 7211-57-6
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/7211-57-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/7211-57-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart