AMBH2D6EF5B7 Ambeed, Inc.

3,4-Dibromo-1-trityl-1H-pyrrole-2,5-dione

  • MDL number MFCD07782026
  • Cas Number : 160989-35-5
  • Molecular Weight : 497.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/160989-35-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/160989-35-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart