AMBH2D6EF5D3 Ambeed, Inc.

Omeprazole sodium

  • MDL number MFCD07802816
  • Cas Number : 95510-70-6
  • Molecular Weight : 367.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/95510-70-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/95510-70-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart