AMBH2D6EF6AA Ambeed, Inc.

3-(Diphenylmethylene)-1,1-diethyl-2-methylpyrrolidin-1-ium bromide

  • MDL number MFCD00865211
  • Cas Number : 4630-95-9
  • Molecular Weight : 386.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-219
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/4630-95-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/4630-95-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart