AMBH2D6EF6DC Ambeed, Inc.

Ethyl 3-amino-1-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD00101268
  • Cas Number : 21230-43-3
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 135.5°C
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 300.5°C at 760 mmHg
Product Link https://www.ambeed.com/products/21230-43-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/21230-43-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart