AMBH2D6EF6E5 Ambeed, Inc.

Pyridin-4-ylmethanamine hydrochloride

  • MDL number MFCD06637470
  • Cas Number : 64460-41-9
  • Molecular Weight : 144.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/64460-41-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/64460-41-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart