AMBH2D6EF6E8 Ambeed, Inc.

2,3-Dichloro-4-methylpyridine

  • MDL number MFCD09839231
  • Cas Number : 191419-07-5
  • Molecular Weight : 162.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 111.2°C
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 224.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/191419-07-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/191419-07-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart