AMBH2D6EF72B Ambeed, Inc.

Ruthenium(III) chloride

  • MDL number MFCD00011208
  • Cas Number : 10049-08-8
  • Molecular Weight : 207.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 500
Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/10049-08-8.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/10049-08-8.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart