AMBH2D6EF733 Ambeed, Inc.

S-Acetyl-L-glutathione

  • MDL number MFCD09952588
  • Cas Number : 3054-47-5
  • Molecular Weight : 349.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(((S)-3-(Acetylthio)-1-((carboxymethyl)amino)-1-oxopropan-2-yl)amino)-2-amino-5-oxopentanoic acid
Purity 95
Flash Point 419.6
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 770.2�t 760 mmHg
Product Link https://www.ambeed.com/products/3054-47-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/3054-47-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart