AMBH2D6EF771 Ambeed, Inc.

Sivelestat sodium tetrahydrate

  • MDL number MFCD01937969
  • Cas Number : 201677-61-4
  • Molecular Weight : 528.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-69
Synonym Sodium 2-(2-(4-(pivaloyloxy)phenylsulfonamido)benzamido)acetate tetrahydrate
Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/201677-61-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/201677-61-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart