AMBH2D6EF785 Ambeed, Inc.

Sodium 2-((2-(stearoyloxy)propanoyl)oxy)propanoate

  • MDL number MFCD00152806
  • Cas Number : 25383-99-7
  • Molecular Weight : 450.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/25383-99-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/25383-99-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart