AMBH2D6EF7AD Ambeed, Inc.

Ethyl 6,7-difluoro-1-methyl-4-oxo-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylate

  • MDL number MFCD08063939
  • Cas Number : 113046-72-3
  • Molecular Weight : 311.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 218
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 436.8�t 760 mmHg
Product Link https://www.ambeed.com/products/113046-72-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/113046-72-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart