AMBH2D6EF7EA Ambeed, Inc.

Tasosartan

  • MDL number MFCD00865905
  • Cas Number : 145733-36-4
  • Molecular Weight : 411.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/145733-36-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/145733-36-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart