AMBH2D6EF80A Ambeed, Inc.

tert-Butyl 2-carbamoylpyrrolidine-1-carboxylate

  • MDL number MFCD02183583
  • Cas Number : 54503-10-5
  • Molecular Weight : 214.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/54503-10-5.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/54503-10-5.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart