AMBH2D6EF887 Ambeed, Inc.

Sodium 4,5-Dihydroxy-3,6-dioxocyclohexa-1,4-diene-1,2-bis(olate)

  • MDL number MFCD00036127
  • Cas Number : 1887-02-1
  • Molecular Weight : 216.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1887-02-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1887-02-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart