AMBH2D6EF88D Ambeed, Inc.

1,3,7,9-Tetramethyl-1H-purine-2,6,8(3H,7H,9H)-trione

  • MDL number MFCD00042781
  • Cas Number : 2309-49-1
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2309-49-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2309-49-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart