AMBH2D6EF8F7 Ambeed, Inc.

(trans,trans)-4'-Pentyl-[1,1'-bi(cyclohexan)]-4-ol

  • MDL number MFCD09752070
  • Cas Number : 82575-70-0
  • Molecular Weight : 252.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/82575-70-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/82575-70-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart