AMBH2D6F014F Ambeed, Inc.

3-Amino-1-methyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD11040163
  • Cas Number : 21230-50-2
  • Molecular Weight : 122.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/21230-50-2.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/21230-50-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart