AMBH2D6F0156 Ambeed, Inc.

1-Methyl-1H-benzo[d]imidazole-6-carbaldehyde

  • MDL number MFCD11977687
  • Cas Number : 181867-19-6
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/181867-19-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/181867-19-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart