AMBH2D6F0192 Ambeed, Inc.

tert-Butyl 4-(3-aminopyrazin-2-yl)piperazine-1-carboxylate

  • MDL number MFCD11040180
  • Cas Number : 1208542-95-3
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1208542-95-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1208542-95-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart