AMBH2D6F01B7 Ambeed, Inc.

2-Phenyl-2,3-dihydroquinolin-4(1H)-one

  • MDL number MFCD00098918
  • Cas Number : 16619-14-0
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 318
Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 410.1�t 760 mmHg
Product Link https://www.ambeed.com/products/16619-14-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/16619-14-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart