AMBH2D6F01F0 Ambeed, Inc.

N,N-Dimethyl-2-(piperazin-1-yl)benzamide oxalate

  • MDL number MFCD11869752
  • Cas Number : 494782-69-3
  • Molecular Weight : 323.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/494782-69-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/494782-69-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart