AMBH2D6F022C Ambeed, Inc.

2-Methyl-1,3,7-triazaspiro[4.5]dec-2-en-4-one hydrochloride

  • MDL number MFCD11869758
  • Cas Number : 1214028-87-1
  • Molecular Weight : 203.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1214028-87-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1214028-87-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart