AMBH2D6F027F Ambeed, Inc.

4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD11518907
  • Cas Number : 1016241-80-7
  • Molecular Weight : 197.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1016241-80-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1016241-80-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart