AMBH2D6F02F9 Ambeed, Inc.

6-Benzyl-6,7-dihydro-1H-pyrrolo[3,4-d]pyrimidine-2,4(3H,5H)-dione

  • MDL number MFCD10699254
  • Cas Number : 635698-34-9
  • Molecular Weight : 243.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/635698-34-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/635698-34-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart