AMBH2D6F034A Ambeed, Inc.

2-(Benzo[d]oxazol-2-ylthio)-4-chlorothiazole-5-carbaldehyde

  • MDL number MFCD03453095
  • Cas Number : 914348-74-6
  • Molecular Weight : 296.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/914348-74-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/914348-74-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart