AMBH2D6F0366 Ambeed, Inc.

tert-Butyl 3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate

  • MDL number MFCD05864712
  • Cas Number : 914349-07-8
  • Molecular Weight : 292.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate
Inchi Code 1S/C14H16N2O5/c1-14(2,3)21-13(18)15-7-9(8-17)11-6-10(16(19)20)4-5-12(11)15/h4-7,17H,8H2,1-3H3
Inchi Key ZMPACHXTNKWGMG-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Sealed in dry,2-8
Physical Form Off white to Pale-yellow Solid or liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/914349-07-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/914349-07-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart