AMBH2D6F0390 Ambeed, Inc.

tert-Butyl 4-methyl-4-(piperazin-1-yl)piperidine-1-carboxylate

  • MDL number MFCD09743693
  • Cas Number : 1185064-24-7
  • Molecular Weight : 283.4100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1185064-24-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1185064-24-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart