AMBH2D6F03B2 Ambeed, Inc.

tert-Butyl 5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

  • MDL number MFCD10566052
  • Cas Number : 873779-30-7
  • Molecular Weight : 381.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/873779-30-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/873779-30-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart