AMBH2D6F03D7 Ambeed, Inc.

5-(Trifluoromethyl)pyrazin-2-amine

  • MDL number MFCD10697797
  • Cas Number : 69816-38-2
  • Molecular Weight : 163.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-133
Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 226.204�t 760 mmHg
Product Link https://www.ambeed.com/products/69816-38-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/69816-38-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart