AMBH2D6F03F2 Ambeed, Inc.

6-Bromochromen-2-one

  • MDL number MFCD01327473
  • Cas Number : 19063-55-9
  • Molecular Weight : 225.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-170
Purity 95
Flash Point 165
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 349.2�t 760 mmHg
Product Link https://www.ambeed.com/products/19063-55-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/19063-55-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart